Assay Details
| Gene Symbol: Slc28a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022317.3 | NP_071712.3 | 8 - 9 | 980 | 89 | |
| GenBank mRNA | AF305211.1 | - | 8 - 9 | 913 | 89 | |
| AK052784.1 | - | 8 - 9 | 981 | 89 | ||
| AK145077.1 | - | 8 - 9 | 981 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc28a3
Genomic Map