Assay Details
| Gene Symbol: Ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026842.4 | NP_081118.4 | 8 - 9 | 1525 | 77 | |
| GenBank mRNA | AK004183.1 | - | 1 - 2 | 809 | 77 | |
| AK049298.1 | - | 8 - 9 | 1280 | 77 | ||
| AK187729.1 | - | 2 - 3 | 139 | 77 | ||
| AK198908.1 | - | 2 - 3 | 139 | 77 | ||
| AK209853.1 | - | 2 - 3 | 139 | 77 | ||
| BC010213.1 | - | 5 - 6 | 868 | 77 | ||
| BC027375.1 | - | 2 - 3 | 228 | 77 | ||
| BC051098.1 | - | 7 - 8 | 1153 | 77 | ||
| BC080667.1 | - | 8 - 9 | 1486 | 4935292 | 77 | |
More Information
Set Membership: |
|

Ubqln1
Genomic Map