Assay Details
| Gene Symbol: 4930470H14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045764.1 | 2 - 3 | 1347 | 153 | ||
| GenBank mRNA | AK015535.1 | - | 2 - 3 | 1349 | 153 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NADPH oxidase 3 | Nox3 |
GP91-3, het, nmf250 |
-375.4kb | Sense | Chr. 17 - 3635240 - 3696261 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930548J01 gene | 4930548J01Rik | 1700113K09Rik | 46.0kb | Sense | Chr. 17 - 4119446 - 4122102 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0100004401.rn
Additional Information:
This assay is in the 5'UTR region of the AK015535.1 transcripts.
Set Membership: |
|

4930470H14Rik
Genomic Map