Assay Details
Gene Symbol: 4930470H14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045764.1 | 3 - 4 | 1432 | 111 | ||
GenBank mRNA | AK015535.1 | - | 3 - 4 | 1434 | 111 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NADPH oxidase 3 | Nox3 |
GP91-3, het, nmf250 |
-348.4kb | Sense | Chr. 17 - 3635240 - 3696261 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930548J01 gene | 4930548J01Rik | 1700113K09Rik | 47.7kb | Sense | Chr. 17 - 4119446 - 4122102 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124926 | AK015535.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124732 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120834 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124732 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120834 | - | - |
Whole Mouse Genome G4122A | A_52_P514990 | AK015535.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121398 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124732 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120834 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009043 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008806 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008996 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008785 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008990 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008795 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008784 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008942 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008963 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13252532 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008987 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008962 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008798 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13252548 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008951 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13348616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008946 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008978 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008952 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008949 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13367601 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008960 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008789 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13575813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008986 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008994 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009046 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13236638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008788 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13367590 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009040 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008792 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008808 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008957 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31009047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008964 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008945 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008944 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008796 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008955 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13334876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31008948 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1454068_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1454068_at | - | - |
Mouse Genome 430 2.0 Array | 1454068_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75815_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75815_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700012325.mm.1 | NR_045764.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700012325.mm.1 | NR_045764.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700012325.mm.1 | TC1700001360.mm.1 | MTA TC |
Mouse Expression Set 430 | 1454068_PM_at | - | - |
Mouse Expression Set 430 | 1454068_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454068_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454068_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 100870097 | - | - |
