Assay Details
Gene Symbol: Olfr1018 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146586.2 | NP_666797.2 | 1 | 513 | 101 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1954226 | NM_146586.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116868 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1954226 | NM_146586.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P420519 | NM_146586.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399505 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399532 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399526 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399531 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399494 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399537 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399535 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399492 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399514 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399541 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399501 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399523 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399528 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399525 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399503 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399538 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399521 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399491 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399498 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399517 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399506 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399488 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399511 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399524 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399512 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399508 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399502 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399493 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399536 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116868 | - | - |
Whole Mouse Genome G4122A | A_51_P420519 | NM_146586.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116868 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01568304 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01568310 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01774542 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01449885 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01774543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399493 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399536 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399484 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399546 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399503 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399514 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399537 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399505 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399555 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01449889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01332852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399493 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399553 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399531 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02190916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399549 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399538 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02246287 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399519 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10473468 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10473467 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1660400 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1660400 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001660400 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232618 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1232618 | - | - |
mouse-6 v1.1 (Illumina) | scl4940.1.1_266-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1660400 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl4940.1.1_266-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660400 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1232618 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232618 | - | - |
