Assay Details
| Gene Symbol: Tfb1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146074.1 | NP_666186.1 | 4 - 5 | 547 | 91 | |
| GenBank mRNA | AF508971.1 | - | 4 - 5 | 547 | 91 | |
| BC032930.1 | - | 4 - 5 | 547 | 1382837 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tfb1m
Genomic Map