| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700000211.mm.1 |
NR_045081.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000211.mm.1 |
NR_045081.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000211.mm.1 |
TC1700000041.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000212.mm.1 |
NR_045081.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000212.mm.1 |
NR_045081.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000212.mm.1 |
TC1700000041.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000217.mm.1 |
NR_045079.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000217.mm.1 |
NR_045079.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000217.mm.1 |
NR_045081.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000217.mm.1 |
NR_045081.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000217.mm.1 |
TC1700000041.mm.1 |
MTA TC |