Assay Details
| Gene Symbol: Lrrc4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289742.1 | NP_001276671.1 | 3 - 4 | 679 | 104 | |
| NM_001289743.1 | NP_001276672.1 | 3 - 4 | 679 | 104 | ||
| GenBank mRNA | AK034276.1 | - | 3 - 4 | 680 | 104 | |
| AK048322.1 | - | 3 - 4 | 633 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034276.1, AK048322.1, NM_001289742.1, NM_001289743.1 transcripts.
Set Membership: |
|

Lrrc4c
Genomic Map