Assay Details
| Gene Symbol: Tmco5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029232.2 | NP_083508.1 | 3 - 4 | 323 | 108 | |
| GenBank mRNA | AK016214.1 | - | 3 - 4 | 314 | 108 | |
| BC050803.1 | - | 3 - 4 | 323 | 6771315 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmco5b
Genomic Map