Assay Details
| Gene Symbol: Tmco5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029232.2 | NP_083508.1 | 7 - 8 | 609 | 85 | |
| GenBank mRNA | AK016214.1 | - | 7 - 8 | 600 | 85 | |
| BC050803.1 | - | 7 - 8 | 609 | 6771315 | 85 | |
More Information
Set Membership: |
|

Tmco5b
Genomic Map