Assay Details
| Gene Symbol: Fmr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290424.1 | NP_001277353.1 | 5 - 6 | 698 | 120 | |
| NM_008031.3 | NP_032057.2 | 5 - 6 | 698 | 120 | ||
| GenBank mRNA | AF461114.1 | - | 5 - 6 | 422 | 120 | |
| AK046605.1 | - | 5 - 6 | 658 | 120 | ||
| AK053211.1 | - | 2 - 3 | 440 | 120 | ||
| AK053701.1 | - | 5 - 6 | 658 | 120 | ||
| AK053829.1 | - | 5 - 6 | 700 | 120 | ||
| AK080948.1 | - | 5 - 6 | 658 | 120 | ||
| AK140343.1 | - | 5 - 6 | 660 | 120 | ||
| BC079671.1 | - | 5 - 6 | 684 | 30532682 | 120 | |
| L23971.1 | - | 5 - 6 | 545 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fmr1
Genomic Map