Assay Details
| Gene Symbol: Fmr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290424.1 | NP_001277353.1 | 10 - 11 | 1263 | 101 | |
| NM_008031.3 | NP_032057.2 | 10 - 11 | 1263 | 101 | ||
| GenBank mRNA | AF461114.1 | - | 10 - 11 | 987 | 101 | |
| AK046605.1 | - | 10 - 11 | 1223 | 101 | ||
| AK053211.1 | - | 7 - 8 | 1005 | 101 | ||
| AK053829.1 | - | 10 - 11 | 1265 | 101 | ||
| AK080948.1 | - | 10 - 11 | 1223 | 101 | ||
| AK140343.1 | - | 10 - 11 | 1225 | 101 | ||
| BC079671.1 | - | 10 - 11 | 1249 | 30532682 | 101 | |
| L23971.1 | - | 10 - 11 | 1110 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fmr1
Genomic Map