Assay Details
| Gene Symbol: Ppp1r13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011625.1 | NP_035755.1 | 13 - 14 | 2908 | 94 | |
| GenBank mRNA | BC034207.1 | - | 9 - 10 | 2354 | 94 | |
| BC053092.1 | - | 13 - 14 | 2934 | 5719383 | 94 | |
| BC053732.1 | - | 9 - 10 | 2354 | 94 | ||
| BC054788.1 | - | 13 - 14 | 2908 | 6826065 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r13b
Genomic Map