Assay Details
| Gene Symbol: Ppp1r13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011625.1 | NP_035755.1 | 9 - 10 | 1337 | 61 | |
| GenBank mRNA | BC053092.1 | - | 9 - 10 | 1363 | 5719383 | 61 |
| BC054788.1 | - | 9 - 10 | 1337 | 6826065 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r13b
Genomic Map