Assay Details
| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111331.1 | NP_001104801.1 | 6 - 7 | 973 | 131 | |
| NM_001291005.1 | NP_001277934.1 | 7 - 8 | 795 | 131 | ||
| NM_019789.4 | NP_062763.2 | 7 - 8 | 799 | 131 | ||
| NR_110989.1 | 7 - 8 | 959 | 131 | |||
| GenBank mRNA | AF274050.1 | - | 6 - 7 | 659 | 131 | |
| AF287732.1 | - | 7 - 8 | 773 | 131 | ||
| AF287733.1 | - | 7 - 8 | 777 | 131 | ||
| AF300870.1 | - | 7 - 8 | 722 | 131 | ||
| AK016721.1 | - | 7 - 8 | 961 | 131 | ||
| AK165711.1 | - | 7 - 8 | 747 | 131 | ||
| BC026980.1 | - | 7 - 8 | 736 | 5368268 | 131 | |
| BC047139.1 | - | 7 - 8 | 736 | 6389719 | 131 | |
| BC057329.1 | - | 7 - 8 | 718 | 6813819 | 131 | |
| DQ148499.1 | - | 6 - 7 | 659 | 131 | ||
| DQ148500.1 | - | 6 - 7 | 973 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnip3
Genomic Map