Assay Details
| Gene Symbol: Fahd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029629.2 | NP_083905.1 | 1 - 2 | 152 | 66 | |
| GenBank mRNA | AK005134.1 | - | 1 - 2 | 153 | 66 | |
| AK080868.1 | - | 1 - 2 | 138 | 66 | ||
| AK170875.1 | - | 1 - 2 | 126 | 66 | ||
| BC030182.1 | - | 1 - 2 | 103 | 5044017 | 66 | |
| BY269533.1 | - | 1 - 2 | 153 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK080868.1, AK170875.1, BC030182.1, NM_029629.2 transcripts.
Set Membership: |
|

Fahd2a
Genomic Map