Assay Details
| Gene Symbol: Slc22a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033167.3 | NP_001028339.1 | 3 - 4 | 1285 | 83 | |
| GenBank mRNA | AK147397.1 | - | 3 - 4 | 1286 | 83 | |
| BC053705.1 | - | 1 - 2 | 73 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a23
Genomic Map