Assay Details
| Gene Symbol: Agps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172666.3 | NP_766254.2 | 17 - 18 | 1768 | 109 | |
| GenBank mRNA | AK031049.1 | - | 17 - 18 | 1770 | 109 | |
| AK031154.1 | - | 17 - 18 | 1609 | 109 | ||
| AK079447.1 | - | 16 - 17 | 1448 | 109 | ||
| AK164922.1 | - | 4 - 5 | 250 | 109 | ||
| BC063086.1 | - | 17 - 18 | 1753 | 6332574 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agps
Genomic Map