Assay Details
| Gene Symbol: D430019H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252508.1 | NP_001239437.1 | 1 - 2 | 297 | 64 | |
| GenBank mRNA | AK052429.1 | - | 1 - 2 | 298 | 64 | |
| AK081193.1 | - | 1 - 2 | 178 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001252508.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P97699 | BC059046.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P97699 | BC078436.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P97699 | AK081558.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P97699 | BC058677.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P97699 | BC059046.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P97699 | BC078436.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P97699 | AK081558.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P97699 | BC058677.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502861 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348735 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648758 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK075973.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK078935.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK088691.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK145191.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK145196.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK145308.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK147787.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK149920.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK150545.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK150553.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK150558.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK150570.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK150662.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK150675.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK150711.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK150879.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151010.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151136.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151145.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151159.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151166.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151190.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151202.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151226.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151277.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151297.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151350.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151385.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151386.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151398.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151444.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151468.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151663.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151898.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151956.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151995.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK151999.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152043.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152052.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152083.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152218.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152433.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152470.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152591.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152615.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152651.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152671.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152813.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152841.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK152844.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK153461.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK159759.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK159834.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK166349.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK167117.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK167825.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK167960.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | AK202772.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | EF095208.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | J04181.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | M12481.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | NM_007393.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | X03672.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608492 | X03765.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903586 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494669 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304225 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802355 | AF507967.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802355 | AY124569.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802355 | BC106861.2 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802355 | BC111522.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802355 | NM_152944.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748007 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209104 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49354 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02597052 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07737194 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02597027 | - | - |
| Whole Mouse Genome G4122A | A_52_P97699 | BC059046.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P97699 | BC078436.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P97699 | AK081558.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P97699 | BC058677.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P97699 | BC059046.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P97699 | BC078436.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P97699 | AK081558.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P97699 | BC058677.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133174 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02597027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10291264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10328651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10314427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09764013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09871585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10259840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10178487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09749357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10250407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09991126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10261349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09770320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10370911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09886443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09836592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10332970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09764017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09886442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09927425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10324340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09764015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10291257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10090136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28709441 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1455447_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455447_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455447_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 268595_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 268595_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200007581.mm.1 | NM_001252508.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007581.mm.1 | NM_001252508.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007581.mm.1 | TC1200001064.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007578.mm.1 | NM_001252508.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007578.mm.1 | NM_001252508.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007578.mm.1 | TC1200001064.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007583.mm.1 | NM_001252508.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007583.mm.1 | NM_001252508.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007583.mm.1 | TC1200001064.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455447_PM_at | - | - |
| Mouse Expression Set 430 | 1455447_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455447_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455447_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455447_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455447_at | - | - |

D430019H16Rik
Genomic Map