Assay Details
| Gene Symbol: Akr1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018859.2 | NP_061347.2 | 3 - 4 | 368 | 79 | |
| GenBank mRNA | AI893997.1 | - | 3 - 4 | 368 | 79 | |
| AK002507.1 | - | 3 - 4 | 343 | 79 | ||
| AK007924.1 | - | 3 - 4 | 345 | 79 | ||
| AK028605.1 | - | 194 | 79 | |||
| BC012692.1 | - | 3 - 4 | 384 | 3986863 | 79 | |
| U68535.1 | - | 3 - 4 | 79 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Akr1e1
Genomic Map