Assay Details
| Gene Symbol: Akr1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018859.2 | NP_061347.2 | 5 - 6 | 567 | 138 | |
| GenBank mRNA | AK002507.1 | - | 5 - 6 | 542 | 138 | |
| AK007924.1 | - | 5 - 6 | 544 | 138 | ||
| AK028605.1 | - | 393 | 138 | |||
| BC012692.1 | - | 5 - 6 | 583 | 3986863 | 138 | |
| U68535.1 | - | 5 - 6 | 553 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akr1e1
Genomic Map