Assay Details
| Gene Symbol: Akr1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018859.2 | NP_061347.2 | 6 - 7 | 662 | 101 | |
| GenBank mRNA | AK002507.1 | - | 6 - 7 | 637 | 101 | |
| AK007924.1 | - | 6 - 7 | 639 | 101 | ||
| AK028605.1 | - | 488 | 101 | |||
| BC012692.1 | - | 6 - 7 | 678 | 3986863 | 101 | |
| U68535.1 | - | 6 - 7 | 648 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akr1e1
Genomic Map