Assay Details
| Gene Symbol: Akr1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018859.2 | NP_061347.2 | 9 - 10 | 908 | 87 | |
| GenBank mRNA | AK002507.1 | - | 9 - 10 | 883 | 87 | |
| AK007924.1 | - | 9 - 10 | 884 | 87 | ||
| AK028605.1 | - | 734 | 87 | |||
| BC012692.1 | - | 9 - 10 | 924 | 3986863 | 87 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043147 | U68535.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043147 | AK028605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043147 | BC012692.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043147 | BQ174229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043147 | NM_018859.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136132 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576893 | AK002688.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576893 | AK012820.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576893 | BC049583.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576893 | NM_025339.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339134 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9359 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106662 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128274 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043147 | U68535.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043147 | AK028605.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043147 | BC012692.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043147 | BQ174229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043147 | NM_018859.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28792911 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128274 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106662 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07772559 | - | - |
| Whole Mouse Genome G4122A | A_52_P496424 | U68535.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496424 | AK028605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496424 | BC012692.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496424 | BQ174229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P496424 | NM_018859.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P195315 | U68535.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133992 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100969 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128274 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106662 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10816739 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10520433 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10520431 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10520437 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10816737 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10520435 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10520441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28792904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11035662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10799065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10799056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10556334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10895876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10656925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28792918 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10407470 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407469 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407478 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407468 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407472 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407474 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407471 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407473 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407479 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407480 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407476 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1446483_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417826_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10407467 | - | - |
| Mouse Genome 430 2.0 Array | 1417826_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446483_at | - | - |
| Mouse Genome 430 2.0 Array | 1417826_at | - | - |
| Mouse Genome 430 2.0 Array | 1458517_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446483_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458517_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56043_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56043_PM_at | - | - |
| Mouse Expression Set 430 | 1417826_at | - | - |
| Mouse Expression Set 430 | 1417826_PM_at | - | - |
| Mouse Expression Set 430 | 1446483_at | - | - |
| Mouse Expression Set 430 | 1446483_PM_at | - | - |
| Murine Genome U74v2 Set | 103068_PM_at | - | - |
| Murine Genome U74v2 Set | 103068_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417826_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417826_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417826_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417826_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446483_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446483_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 838998 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3360736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3360736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2602837 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2602837 | - | - |
| mouse-6 v1.1 (Illumina) | scl056043.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056043.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2602837 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2602837 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl056043.1_7-S | - | - |

Akr1e1
Genomic Map