Assay Details
| Gene Symbol: Actn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033268.4 | NP_150371.4 | 10 - 11 | 1341 | 79 | |
| GenBank mRNA | AY036877.1 | - | 11 - 12 | 1285 | 79 | |
| BC089579.1 | - | 10 - 11 | 1341 | 6757718 | 79 | |
More Information
Set Membership: |
|

Actn2
Genomic Map