| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200035357.mm.1 |
NM_020267.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035357.mm.1 |
NM_020267.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035357.mm.1 |
TC0200004213.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035358.mm.1 |
NM_020267.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035358.mm.1 |
NM_020267.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035358.mm.1 |
TC0200004213.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035356.mm.1 |
NM_020267.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035356.mm.1 |
NM_020267.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035356.mm.1 |
TC0200004213.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035359.mm.1 |
NM_020267.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035359.mm.1 |
NM_020267.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035359.mm.1 |
TC0200004213.mm.1 |
MTA TC |