Assay Details
| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020267.2 | NP_064663.2 | 2 - 3 | 965 | 135 | |
| GenBank mRNA | AJ249129.1 | - | 2 - 3 | 914 | 135 | |
| AJ278191.1 | - | 2 - 3 | 973 | 135 | ||
| BC039979.1 | - | 2 - 3 | 391 | 135 | ||
| BC045602.1 | - | 2 - 3 | 380 | 135 | ||
| BC098369.1 | - | 2 - 3 | 934 | 6830438 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim44
Genomic Map