Assay Details
| Gene Symbol: Apip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019735.4 | NP_062709.3 | 3 - 4 | 316 | 69 | |
| GenBank mRNA | AK077705.1 | - | 3 - 4 | 317 | 69 | |
| AK145450.1 | - | 3 - 4 | 318 | 69 | ||
| BC028434.1 | - | 3 - 4 | 259 | 1245515 | 69 | |
More Information
Set Membership: |
|

Apip
Genomic Map