Assay Details
| Gene Symbol: Lypd6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027990.3 | NP_082266.1 | 7 - 8 | 633 | 109 | |
| GenBank mRNA | AK009282.1 | - | 7 - 8 | 633 | 109 | |
| BC126943.1 | - | 4 - 5 | 377 | 40090023 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lypd6b
Genomic Map