Assay Details
| Gene Symbol: Spag6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001334.2 | NP_001001334.2 | 5 - 6 | 723 | 69 | |
| GenBank mRNA | AK132882.1 | - | 5 - 6 | 723 | 69 | |
| BC061194.1 | - | 5 - 6 | 722 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spag6l
Genomic Map