Assay Details
| Gene Symbol: Fbxo18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015792.1 | NP_056607.1 | 11 - 12 | 1965 | 87 | |
| GenBank mRNA | AF184275.1 | - | 9 - 10 | 1459 | 87 | |
| AK083118.1 | - | 11 - 12 | 2310 | 87 | ||
| AK084645.1 | - | 11 - 12 | 1901 | 87 | ||
| AK147829.1 | - | 11 - 12 | 1949 | 87 | ||
| AK154357.1 | - | 11 - 12 | 1988 | 87 | ||
| AK158937.1 | - | 11 - 12 | 2016 | 87 | ||
| BC004036.1 | - | 8 - 9 | 1051 | 87 | ||
| BC031393.1 | - | 11 - 12 | 1965 | 4948804 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo18
Genomic Map