Assay Details
| Gene Symbol: Fbxo18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015792.1 | NP_056607.1 | 5 - 6 | 1108 | 58 | |
| GenBank mRNA | AF184275.1 | - | 3 - 4 | 602 | 58 | |
| AK083118.1 | - | 5 - 6 | 1453 | 58 | ||
| AK084645.1 | - | 5 - 6 | 1044 | 58 | ||
| AK147829.1 | - | 5 - 6 | 1092 | 58 | ||
| AK154357.1 | - | 5 - 6 | 1131 | 58 | ||
| AK158937.1 | - | 5 - 6 | 1159 | 58 | ||
| BC004036.1 | - | 2 - 3 | 194 | 58 | ||
| BC031393.1 | - | 5 - 6 | 1108 | 4948804 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo18
Genomic Map