Assay Details
| Gene Symbol: Pfkfb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177752.1 | NP_001171223.1 | 1 - 2 | 464 | 88 | |
| NM_001177753.1 | NP_001171224.1 | 1 - 2 | 464 | 88 | ||
| NM_001177754.1 | NP_001171225.1 | 1 - 2 | 464 | 88 | ||
| NM_001177755.1 | NP_001171226.1 | 1 - 2 | 464 | 88 | ||
| NM_001177756.1 | NP_001171227.1 | 1 - 2 | 464 | 88 | ||
| NM_001177757.1 | NP_001171228.1 | 1 - 2 | 464 | 88 | ||
| NM_133232.3 | NP_573495.2 | 1 - 2 | 464 | 88 | ||
| GenBank mRNA | AF294617.1 | - | 1 - 2 | 399 | 88 | |
| AK054285.1 | - | 1 - 2 | 449 | 88 | ||
| AK154607.1 | - | 1 - 2 | 414 | 88 | ||
| BC052400.1 | - | 1 - 2 | 441 | 5721049 | 88 | |
| BC057320.1 | - | 1 - 2 | 416 | 6826100 | 88 | |
| EU034675.1 | - | 1 - 2 | 74 | 88 | ||
| EU034676.1 | - | 1 - 2 | 74 | 88 | ||
| EU034677.1 | - | 1 - 2 | 74 | 88 | ||
| EU034678.1 | - | 1 - 2 | 74 | 88 | ||
| EU034679.1 | - | 1 - 2 | 74 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkfb3
Genomic Map