Assay Details
| Gene Symbol: Pfkfb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177752.1 | NP_001171223.1 | 3 - 4 | 696 | 83 | |
| NM_001177753.1 | NP_001171224.1 | 3 - 4 | 696 | 83 | ||
| NM_001177754.1 | NP_001171225.1 | 3 - 4 | 696 | 83 | ||
| NM_001177755.1 | NP_001171226.1 | 3 - 4 | 696 | 83 | ||
| NM_001177756.1 | NP_001171227.1 | 3 - 4 | 696 | 83 | ||
| NM_001177757.1 | NP_001171228.1 | 3 - 4 | 696 | 83 | ||
| NM_001177758.1 | NP_001171229.1 | 3 - 4 | 333 | 83 | ||
| NM_133232.3 | NP_573495.2 | 3 - 4 | 696 | 83 | ||
| GenBank mRNA | AF294617.1 | - | 3 - 4 | 631 | 83 | |
| AK054285.1 | - | 3 - 4 | 681 | 83 | ||
| AK077210.1 | - | 3 - 4 | 282 | 83 | ||
| AK138807.1 | - | 3 - 4 | 481 | 83 | ||
| AK143330.1 | - | 3 - 4 | 303 | 83 | ||
| AK154607.1 | - | 3 - 4 | 646 | 83 | ||
| BC052400.1 | - | 3 - 4 | 673 | 5721049 | 83 | |
| BC057320.1 | - | 3 - 4 | 648 | 6826100 | 83 | |
| EU034675.1 | - | 3 - 4 | 306 | 83 | ||
| EU034676.1 | - | 3 - 4 | 306 | 83 | ||
| EU034677.1 | - | 3 - 4 | 306 | 83 | ||
| EU034678.1 | - | 3 - 4 | 306 | 83 | ||
| EU034679.1 | - | 3 - 4 | 306 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkfb3
Genomic Map