Assay Details
| Gene Symbol: Prkcq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008859.2 | NP_032885.1 | 10 - 11 | 1101 | 83 | |
| GenBank mRNA | AB062122.1 | - | 3 - 4 | 499 | 83 | |
| AK037664.1 | - | 10 - 11 | 1173 | 83 | ||
| AK085546.1 | - | 10 - 11 | 1371 | 83 | ||
| BC138552.1 | - | 9 - 10 | 1032 | 8861572 | 83 | |
| BC145769.1 | - | 9 - 10 | 1032 | 40131072 | 83 | |
| D11091.1 | - | 10 - 11 | 1101 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkcq
Genomic Map