Assay Details
| Gene Symbol: Prkcq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008859.2 | NP_032885.1 | 12 - 13 | 1444 | 85 | |
| GenBank mRNA | AB062122.1 | - | 5 - 6 | 842 | 85 | |
| AK037664.1 | - | 12 - 13 | 1516 | 85 | ||
| BC138552.1 | - | 11 - 12 | 1375 | 8861572 | 85 | |
| BC145769.1 | - | 11 - 12 | 1375 | 40131072 | 85 | |
| D11091.1 | - | 12 - 13 | 1444 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkcq
Genomic Map