| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_001111289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_001111289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_001111290.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_001111290.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_001111291.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_001111291.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_001111292.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_001111292.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_016739.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
NM_016739.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035588.mm.1 |
TC0200004229.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035589.mm.1 |
NM_001111289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035589.mm.1 |
NM_001111289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035589.mm.1 |
NM_001111291.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035589.mm.1 |
NM_001111291.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035589.mm.1 |
NM_001111292.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035589.mm.1 |
NM_001111292.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035589.mm.1 |
NM_016739.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035589.mm.1 |
NM_016739.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035589.mm.1 |
TC0200004229.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_001111289.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_001111289.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_001111290.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_001111290.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_001111291.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_001111291.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_001111292.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_001111292.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_016739.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
NM_016739.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200035591.mm.1 |
TC0200004229.mm.1 |
MTA TC |