Assay Details
| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009804.2 | NP_033934.2 | 7 - 8 | 1013 | 83 | |
| GenBank mRNA | AK075853.1 | - | 7 - 8 | 983 | 83 | |
| AK088168.1 | - | 7 - 8 | 975 | 83 | ||
| AK128919.1 | - | 7 - 8 | 988 | 83 | ||
| AK133881.1 | - | 7 - 8 | 981 | 83 | ||
| AK148961.1 | - | 7 - 8 | 981 | 83 | ||
| AK150893.1 | - | 7 - 8 | 984 | 83 | ||
| AK159152.1 | - | 7 - 8 | 984 | 83 | ||
| AK159885.1 | - | 7 - 8 | 984 | 83 | ||
| AK159891.1 | - | 7 - 8 | 935 | 83 | ||
| AK159913.1 | - | 7 - 8 | 965 | 83 | ||
| AK169069.1 | - | 7 - 8 | 978 | 83 | ||
| AY040626.1 | - | 7 - 8 | 954 | 83 | ||
| BC013447.1 | - | 7 - 8 | 970 | 4241333 | 83 | |
| BC047126.1 | - | 7 - 8 | 928 | 83 | ||
| L25069.1 | - | 7 - 8 | 985 | 83 | ||
| M62897.1 | - | 7 - 8 | 898 | 83 | ||
| X52108.1 | - | 7 - 8 | 898 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cat
Genomic Map