Assay Details
| Gene Symbol: Mrpl36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053163.1 | NP_444393.1 | 2 | 567 | 88 | |
| GenBank mRNA | AK131597.1 | - | 2 | 559 | 88 | |
| AK131601.1 | - | 2 | 557 | 88 | ||
| AK131604.1 | - | 2 | 556 | 88 | ||
| AK160778.1 | - | 2 | 568 | 88 | ||
| BC009166.1 | - | 2 | 567 | 2811744 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139148 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139151 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139152 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139150 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | AK160778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | AK131601.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | BC009166.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | NM_053163.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | AK131597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | AK160778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | AK131601.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | BC009166.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | NM_053163.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450123 | AK131597.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816094 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135329 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | AK160778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | AK131601.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | BC009166.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | NM_053163.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | AK131597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | AK160778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | AK131601.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | BC009166.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | NM_053163.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450123 | AK131597.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | AK160778.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | AK131601.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | BC009166.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | NM_053163.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | AK131597.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | AK160778.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | AK131601.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | BC009166.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | NM_053163.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P450123 | AK131597.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29139151 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135329 | - | - |
| Whole Mouse Genome G4122A | A_51_P450123 | AK160778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450123 | AK131601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450123 | BC009166.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450123 | NM_053163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450123 | AK131597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450123 | AK160778.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450123 | AK131601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450123 | BC009166.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450123 | NM_053163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P450123 | AK131597.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135329 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10969734 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139150 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139152 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139151 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29139148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29139150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10882575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10969732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10516511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10467628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10516510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10919217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10516504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10525702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10766125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10525706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10611403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10525695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10969725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10525688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10790258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10969721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10790259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10516514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29139150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10516503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10406030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406027 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10406028 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422819_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10406026 | - | - |
| Mouse Genome 430 2.0 Array | 1422819_at | - | - |
| Mouse Genome 430 2.0 Array | 1422819_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94066_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 94066_PM_at | - | - |
| Mouse Expression Set 430 | 1422819_at | - | - |
| Mouse Expression Set 430 | 1422819_PM_at | - | - |
| Murine Genome U74v2 Set | 96098_at | - | - |
| Murine Genome U74v2 Set | 96098_PM_at | - | - |
| Murine Genome U74v2 Set | 161488_PM_r_at | - | - |
| Murine Genome U74v2 Set | 161488_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422819_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422819_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422819_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422819_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 442221 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1450048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259906 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259906 | - | - |
| mouse-6 v1.1 (Illumina) | scl44635.2.1_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44635.2.1_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450048 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2797212 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2797212 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl44635.2.1_29-S | - | - |

Mrpl36
Genomic Map