Assay Details
| Gene Symbol: Lpcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145376.5 | NP_663351.3 | 12 - 13 | 1275 | 61 | |
| GenBank mRNA | AB244717.2 | - | 12 - 13 | 1275 | 61 | |
| AK046079.1 | - | 10 - 11 | 1070 | 61 | ||
| AK046507.1 | - | 10 - 11 | 1070 | 61 | ||
| AK081865.1 | - | 12 - 13 | 1433 | 61 | ||
| AK132379.1 | - | 12 - 13 | 1409 | 61 | ||
| AK155286.1 | - | 12 - 13 | 1439 | 61 | ||
| AK169190.1 | - | 12 - 13 | 1408 | 61 | ||
| AK170723.1 | - | 12 - 13 | 1428 | 61 | ||
| AK171582.1 | - | 12 - 13 | 1428 | 61 | ||
| BC005662.1 | - | 10 - 11 | 805 | 61 | ||
| BC066809.1 | - | 12 - 13 | 1224 | 30475544 | 61 | |
More Information
Set Membership: |
|

Lpcat1
Genomic Map