Assay Details
| Gene Symbol: Lpcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145376.5 | NP_663351.3 | 3 - 4 | 487 | 65 | |
| GenBank mRNA | AB244717.2 | - | 3 - 4 | 487 | 65 | |
| AK079946.1 | - | 645 | 65 | |||
| AK081865.1 | - | 3 - 4 | 645 | 65 | ||
| AK132379.1 | - | 3 - 4 | 621 | 65 | ||
| AK155286.1 | - | 3 - 4 | 651 | 65 | ||
| AK169190.1 | - | 3 - 4 | 620 | 65 | ||
| AK171582.1 | - | 3 - 4 | 640 | 65 | ||
| BC066809.1 | - | 3 - 4 | 436 | 30475544 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK079946.1 transcripts.
Set Membership: |
|

Lpcat1
Genomic Map