Assay Details
| Gene Symbol: Unc13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029873.2 | NP_001025044.2 | 27 - 28 | 3432 | 83 | |
| GenBank mRNA | AK147408.1 | - | 22 - 23 | 2945 | 83 | |
| AY753536.1 | - | 27 - 28 | 3322 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Unc13a
Genomic Map