Assay Details
| Gene Symbol: Unc13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029873.2 | NP_001025044.2 | 38 - 39 | 4408 | 68 | |
| GenBank mRNA | AK147408.1 | - | 33 - 34 | 3921 | 68 | |
| AY753536.1 | - | 38 - 39 | 4298 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Unc13a
Genomic Map