Assay Details
| Gene Symbol: Unc13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029873.2 | NP_001025044.2 | 8 - 9 | 665 | 68 | |
| GenBank mRNA | AK010728.1 | - | 7 - 8 | 616 | 68 | |
| AK147408.1 | - | 3 - 4 | 178 | 68 | ||
| AK170263.1 | - | 6 - 7 | 2264 | 68 | ||
| AY753536.1 | - | 8 - 9 | 555 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Unc13a
Genomic Map