Assay Details
| Gene Symbol: Use1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145780.1 | NP_001139252.1 | 5 - 6 | 647 | 69 | |
| NM_025917.4 | NP_080193.1 | 4 - 5 | 434 | 69 | ||
| GenBank mRNA | AF353245.1 | - | 4 - 5 | 427 | 69 | |
| AK003257.1 | - | 4 - 5 | 427 | 69 | ||
| AK003385.1 | - | 3 - 4 | 298 | 69 | ||
| AK008573.1 | - | 4 - 5 | 427 | 69 | ||
| AK008661.1 | - | 3 - 4 | 332 | 69 | ||
| AK012490.1 | - | 4 - 5 | 428 | 69 | ||
| AK017484.1 | - | 2 - 3 | 247 | 69 | ||
| AK160726.1 | - | 4 - 5 | 427 | 69 | ||
| AK165639.1 | - | 4 - 5 | 576 | 69 | ||
| AY842453.1 | - | 4 - 5 | 413 | 69 | ||
| BC025086.1 | - | 3 - 4 | 510 | 3995001 | 69 | |
| BC038519.1 | - | 3 - 4 | 494 | 1428854 | 69 | |
| BC075695.1 | - | 5 - 6 | 553 | 30623689 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017484.1 transcripts.
Set Membership: |
|

Use1
Genomic Map