Assay Details
| Gene Symbol: Use1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145780.1 | NP_001139252.1 | 6 - 7 | 662 | 61 | |
| NM_025917.4 | NP_080193.1 | 5 - 6 | 449 | 61 | ||
| GenBank mRNA | AF353245.1 | - | 5 - 6 | 442 | 61 | |
| AK003257.1 | - | 5 - 6 | 442 | 61 | ||
| AK003385.1 | - | 4 - 5 | 313 | 61 | ||
| AK008573.1 | - | 5 - 6 | 442 | 61 | ||
| AK008661.1 | - | 4 - 5 | 347 | 61 | ||
| AK012490.1 | - | 5 - 6 | 443 | 61 | ||
| AK160726.1 | - | 5 - 6 | 442 | 61 | ||
| AK165639.1 | - | 5 - 6 | 591 | 61 | ||
| AY842453.1 | - | 5 - 6 | 428 | 61 | ||
| BC075695.1 | - | 6 - 7 | 568 | 30623689 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Use1
Genomic Map