Assay Details
| Gene Symbol: Pak1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026550.3 | NP_080826.2 | 5 - 6 | 823 | 132 | |
| GenBank mRNA | AF386076.1 | - | 5 - 6 | 809 | 132 | |
| AK002852.1 | - | 5 - 6 | 573 | 132 | ||
| AK077846.1 | - | 5 - 6 | 811 | 132 | ||
| AK077941.1 | - | 5 - 6 | 570 | 132 | ||
| AK135946.1 | - | 5 - 6 | 570 | 132 | ||
| AK150747.1 | - | 5 - 6 | 571 | 132 | ||
| AK150919.1 | - | 5 - 6 | 571 | 132 | ||
| AK189086.1 | - | 4 - 5 | 331 | 132 | ||
| BC018522.1 | - | 5 - 6 | 566 | 132 | ||
| BC043461.1 | - | 5 - 6 | 739 | 132 | ||
| BC043725.1 | - | 5 - 6 | 739 | 132 | ||
| BC116729.1 | - | 5 - 6 | 554 | 132 | ||
| BC116731.1 | - | 5 - 6 | 554 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1ip1
Genomic Map