Assay Details
| Gene Symbol: Pak1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026550.3 | NP_080826.2 | 7 - 8 | 1057 | 110 | |
| GenBank mRNA | AF386076.1 | - | 7 - 8 | 1043 | 110 | |
| AK002852.1 | - | 7 - 8 | 807 | 110 | ||
| AK077846.1 | - | 7 - 8 | 1045 | 110 | ||
| AK077941.1 | - | 7 - 8 | 804 | 110 | ||
| AK135946.1 | - | 7 - 8 | 804 | 110 | ||
| AK150747.1 | - | 7 - 8 | 805 | 110 | ||
| AK150919.1 | - | 7 - 8 | 805 | 110 | ||
| AY030406.1 | - | 7 - 8 | 872 | 110 | ||
| BC043461.1 | - | 7 - 8 | 973 | 110 | ||
| BC043725.1 | - | 7 - 8 | 973 | 110 | ||
| BC060371.1 | - | 6 - 7 | 1081 | 110 | ||
| BC116729.1 | - | 7 - 8 | 788 | 110 | ||
| BC116731.1 | - | 7 - 8 | 788 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1ip1
Genomic Map