Assay Details
| Gene Symbol: Sap30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021788.2 | NP_068560.1 | 3 - 4 | 840 | 134 | |
| GenBank mRNA | AF075136.1 | - | 3 - 4 | 851 | 134 | |
| AK010928.1 | - | 3 - 4 | 841 | 134 | ||
| AK088745.1 | - | 3 - 4 | 845 | 134 | ||
| BC132081.1 | - | 3 - 4 | 823 | 40130358 | 134 | |
| BC132087.1 | - | 3 - 4 | 823 | 40130364 | 134 | |
| CO800093.1 | - | 1 - 2 | 134 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
skeletal muscle cell differentiation
negative regulation of nucleic acid-templated transcription
DNA binding
transcription corepressor activity
protein binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
skeletal muscle cell differentiation
negative regulation of nucleic acid-templated transcription
DNA binding
transcription corepressor activity
protein binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891493 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891488 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891491 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891492 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891485 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891486 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891494 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891490 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | AK088745.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | BC132081.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | BC132087.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | CO800093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | NM_021788.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | AK088745.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | BC132081.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | BC132087.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | CO800093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432199 | NM_021788.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852974 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129616 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | AK088745.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | BC132081.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | BC132087.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | CO800093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | NM_021788.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | AK088745.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | BC132081.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | BC132087.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | CO800093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432199 | NM_021788.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | AK088745.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | BC132081.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | BC132087.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | CO800093.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | NM_021788.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | AK088745.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | BC132081.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | BC132087.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | CO800093.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P432199 | NM_021788.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891474 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129616 | - | - |
| Whole Mouse Genome G4122A | A_51_P432199 | AK088745.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432199 | BC132081.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432199 | BC132087.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432199 | CO800093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432199 | NM_021788.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432199 | AK088745.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432199 | BC132081.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432199 | BC132087.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432199 | CO800093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432199 | NM_021788.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117594 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101744 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129616 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06928351 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06928349 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07135024 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06928348 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06928346 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891486 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891485 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891492 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891491 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891488 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891494 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891490 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06836769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06983745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06983753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06671940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06928349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07190009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06928351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06983744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06671937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06928350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06983749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07417946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891493 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10578767 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10578766 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10578764 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10578765 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417719_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10578763 | - | - |
| Mouse Genome 430 2.0 Array | 1417719_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417719_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 60406_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 60406_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800017165.mm.1 | NM_021788.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800017165.mm.1 | NM_021788.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800017165.mm.1 | TC0800002252.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800017166.mm.1 | NM_021788.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800017166.mm.1 | NM_021788.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800017166.mm.1 | TC0800002252.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417719_at | - | - |
| Mouse Expression Set 430 | 1417719_PM_at | - | - |
| Murine Genome U74v2 Set | 160068_at | - | - |
| Murine Genome U74v2 Set | 160068_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417719_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417719_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417719_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417719_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5080735 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5080735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005080735 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694917 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2694917 | - | - |
| mouse-6 v1.1 (Illumina) | scl34795.4.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5080735 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34795.4.1_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080735 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2694917 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694917 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34795.4.1_9-S | - | - |

Sap30
Genomic Map