Assay Details
| Gene Symbol: Galnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144731.4 | NP_653332.3 | 5 - 6 | 1040 | 92 | |
| GenBank mRNA | AF349573.1 | - | 5 - 6 | 964 | 92 | |
| AK030683.1 | - | 5 - 6 | 988 | 92 | ||
| AK033427.1 | - | 5 - 6 | 989 | 92 | ||
| AK151424.1 | - | 4 - 5 | 631 | 92 | ||
| AK151753.1 | - | 4 - 5 | 631 | 92 | ||
| AK153143.1 | - | 4 - 5 | 631 | 92 | ||
| BC007484.1 | - | 1 - 2 | 78 | 92 | ||
| BC049907.1 | - | 1 - 2 | 87 | 92 | ||
| BC052461.1 | - | 5 - 6 | 1016 | 6834581 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt7
Genomic Map