Assay Details
| Gene Symbol: Nox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172203.2 | NP_757340.1 | 10 - 11 | 1439 | 67 | |
| GenBank mRNA | AF539799.1 | - | 10 - 11 | 1293 | 67 | |
| AK136432.1 | - | 10 - 11 | 1354 | 67 | ||
| AY174116.1 | - | 7 - 8 | 1034 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nox1
Genomic Map